ENA024294667 Enamine

5-chloro-1-[(2,4-dichlorophenyl)methyl]-3-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD05263118
  • Cas Number : 956713-96-5
  • Molecular Weight : 319.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-08628

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08628
Availability details for
Quantity
  • Availability
  • Add to Cart