ENA024294672 Enamine

2-(2-nitrobenzenesulfonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

  • MDL number MFCD06336085
  • Cas Number : 1008002-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-08633

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08633
Availability details for
Quantity
  • Availability
  • Add to Cart