ENA024294679 Enamine

1-(anilinocarbonyl)piperidine-4-carboxylic acid

  • MDL number MFCD03015374
  • Cas Number : 147636-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178...183
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08640

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08640
Availability details for
Quantity
  • Availability
  • Add to Cart