ENA024294683 Enamine

5-methyl-4-(2,4,5-trimethylphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD05263192
  • Cas Number : 743444-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08644

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08644
Availability details for
Quantity
  • Availability
  • Add to Cart