ENA024294711 Enamine

3-(7-butyl-2,6-dioxo-3-propyl-2,3,6,7-tetrahydro-1H-purin-8-yl)propanoic acid

  • MDL number MFCD05262951
  • Cas Number : 743441-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08672
Availability details for
Quantity
  • Availability
  • Add to Cart