ENA024294714 Enamine

2-{4-[3-(trifluoromethyl)benzenesulfonamido]phenyl}acetic acid

  • MDL number MFCD04614042
  • Cas Number : 735305-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-109
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08675

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08675
Availability details for
Quantity
  • Availability
  • Add to Cart