ENA024294728 Enamine

3-[(2-morpholin-4-ylethyl)amino]propanenitrile

  • MDL number MFCD04623526
  • Cas Number : 41832-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08689

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08689
Availability details for
Quantity
  • Availability
  • Add to Cart