ENA024294737 Enamine

2-(piperidin-1-yl)-4,5-dihydro-1,3-thiazol-4-one

  • MDL number MFCD00763752
  • Cas Number : 31101-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-86
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08698

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08698
Availability details for
Quantity
  • Availability
  • Add to Cart