ENA024294739 Enamine

6-methylindolo[1,2-c]quinazolin-12-amine

  • MDL number MFCD04623039
  • Cas Number : 55338-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-121
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-08700

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08700
Availability details for
Quantity
  • Availability
  • Add to Cart