ENA024294745 Enamine

N-[(5-chloro-1,3-benzoxazol-2-yl)methylidene]hydroxylamine

  • Cas Number : 27412-06-2
  • Molecular Weight : 196.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-151
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08706

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08706
Availability details for
Quantity
  • Availability
  • Add to Cart