ENA024294752 Enamine

2-morpholin-4-yl-5-(piperidin-1-ylsulfonyl)aniline

  • MDL number MFCD04623646
  • Cas Number : 53162-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08713

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08713
Availability details for
Quantity
  • Availability
  • Add to Cart