ENA024294756 Enamine

5-(3-nitrophenyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD00469759

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 253-254
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08717

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08717
Availability details for
Quantity
  • Availability
  • Add to Cart