ENA024294777 Enamine

{[4-(2-methoxyphenyl)-5-methyl-4H-1,2,4-triazol-3-yl]thio}acetic acid

  • MDL number MFCD04625436
  • Cas Number : 731001-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08738

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08738
Availability details for
Quantity
  • Availability
  • Add to Cart