ENA024294780 Enamine

2-chloro-1-[1-(2,3-dihydro-1,4-benzodioxin-2-ylmethyl)-2,5-dimethyl-1H-pyrrol-3-yl]ethanone

  • MDL number MFCD04625438
  • Cas Number : 731002-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08741

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08741
Availability details for
Quantity
  • Availability
  • Add to Cart