ENA024294792 Enamine

2-chloro-1-[5-(furan-2-yl)-3-(4-methylphenyl)-4,5-dihydro-1H-pyrazol-1-yl]ethan-1-one

  • MDL number MFCD04625492
  • Cas Number : 625109-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-08753

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08753
Availability details for
Quantity
  • Availability
  • Add to Cart