ENA024294800 Enamine

1-allyl-5-(5-fluoro-2-hydroxybenzoyl)-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD03984065
  • Cas Number : 737820-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08761

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08761
Availability details for
Quantity
  • Availability
  • Add to Cart