ENA024294814 Enamine

6-amino-4-(4-fluorophenyl)-1-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD04635881
  • Cas Number : 733030-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08775

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08775
Availability details for
Quantity
  • Availability
  • Add to Cart