ENA024294819 Enamine

3-chloro-N-(3-methyl-1-phenyl-1H-pyrazol-5-yl)quinoxalin-2-amine

  • MDL number MFCD02709749
  • Cas Number : 376364-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08780

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08780
Availability details for
Quantity
  • Availability
  • Add to Cart