ENA024294821 Enamine

benzylhydrazine

  • MDL number MFCD00869518
  • Cas Number : 555-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143...145
Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08782

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08782
Availability details for
Quantity
  • Availability
  • Add to Cart