ENA024294831 Enamine

2-[(furan-2-yl)formamido]-4-(methylsulfanyl)butanoic acid

  • MDL number MFCD00573255
  • Cas Number : 65054-95-7
  • Molecular Weight : 243.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-08792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08792
Availability details for
Quantity
  • Availability
  • Add to Cart