ENA024294834 Enamine

butylhydrazine

  • MDL number MFCD06655022

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154...158
Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08795

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08795
Availability details for
Quantity
  • Availability
  • Add to Cart