ENA024294842 Enamine

4-(4-chlorophenyl)-5-[1-(dimethylamino)ethyl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD06335058
  • Cas Number : 750599-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08803

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08803
Availability details for
Quantity
  • Availability
  • Add to Cart