ENA024294843 Enamine

5-[1-(dimethylamino)propyl]-4-[2-(morpholin-4-yl)ethyl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD06660496
  • Cas Number : 879362-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08804

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08804
Availability details for
Quantity
  • Availability
  • Add to Cart