ENA024294848 Enamine

5-[1-(dimethylamino)propyl]-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD06336091
  • Cas Number : 749906-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08809

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08809
Availability details for
Quantity
  • Availability
  • Add to Cart