ENA024294851 Enamine

2-[(1,3-benzothiazol-2-ylthio)methyl]benzoic acid

  • MDL number MFCD04614082
  • Cas Number : 100961-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08812

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08812
Availability details for
Quantity
  • Availability
  • Add to Cart