ENA024294857 Enamine

2-[(phenylcarbamoyl)amino]propanoic acid

  • MDL number MFCD00483853
  • Cas Number : 33653-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-181
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08818

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08818
Availability details for
Quantity
  • Availability
  • Add to Cart