ENA024294858 Enamine

3-[(phenylcarbamoyl)amino]propanoic acid

  • MDL number MFCD00099344
  • Cas Number : 10250-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-139
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08819

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08819
Availability details for
Quantity
  • Availability
  • Add to Cart