ENA024294862 Enamine

4-{2-[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]acetamido}benzoic acid

  • MDL number MFCD03640799
  • Cas Number : 712344-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-191
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08823

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08823
Availability details for
Quantity
  • Availability
  • Add to Cart