ENA024294877 Enamine

3-hydroxy-N-pyridin-2-yl-2-naphthamide

  • MDL number MFCD06335042
  • Cas Number : 24445-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236...238
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08838

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08838
Availability details for
Quantity
  • Availability
  • Add to Cart