ENA024294885 Enamine

2-[(4-acetyl-2-fluorophenyl)sulfanyl]acetic acid

  • MDL number MFCD06336089
  • Cas Number : 749906-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-141
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08846
Availability details for
Quantity
  • Availability
  • Add to Cart