ENA024294893 Enamine

2-cyano-N-[4-(pyridin-4-yl)-1,3-thiazol-2-yl]acetamide

  • MDL number MFCD06335073
  • Cas Number : 750599-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08854
Availability details for
Quantity
  • Availability
  • Add to Cart