ENA024294902 Enamine

5-{[2-(3,4-dimethoxyphenyl)ethyl]amino}-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD05261229
  • Cas Number : 742094-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159-160
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08863

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08863
Availability details for
Quantity
  • Availability
  • Add to Cart