ENA024294905 Enamine

1,1-diethylthiourea

  • MDL number MFCD00463355
  • Cas Number : 7204-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99-101
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08866

Safety Information

signalword Danger
pcodes P264,P270,P301+P310,P330,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-08866
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart