ENA024294908 Enamine

1-(4-fluorophenyl)-2-(methylsulfanyl)-1H-imidazole-5-carboxylic acid

  • MDL number MFCD06335024
  • Cas Number : 750598-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08869
Availability details for
Quantity
  • Availability
  • Add to Cart