ENA024294917 Enamine

4-(5-tert-butyl-2-methylbenzenesulfonamido)butanoic acid

  • MDL number MFCD04637793
  • Cas Number : 735297-18-4
  • Molecular Weight : 313.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-{[(5-tert-butyl-2-methylphenyl)sulfonyl]amino}butanoic acid
Inchi Code 1S/C15H23NO4S/c1-11-7-8-12(15(2,3)4)10-13(11)21(19,20)16-9-5-6-14(17)18/h7-8,10,16H,5-6,9H2,1-4H3,(H,17,18)
Inchi Key XUNXKZAKYDYDSX-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-08878

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08878
Availability details for
Quantity
  • Availability
  • Add to Cart