ENA024294923 Enamine

3-[(1,2,3,4-tetrahydronaphthalen-1-ylamino)sulfonyl]benzoic acid

  • MDL number MFCD04631935
  • Cas Number : 731797-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08884

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08884
Availability details for
Quantity
  • Availability
  • Add to Cart