ENA024294932 Enamine

2-chloro-4-methyl-8-(propan-2-yl)quinoline

  • MDL number MFCD06336088
  • Cas Number : 749906-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08893

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08893
Availability details for
Quantity
  • Availability
  • Add to Cart