ENA024294942 Enamine

3-(prop-2-en-1-yloxy)benzaldehyde

  • MDL number MFCD04227161
  • Cas Number : 40359-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-08903

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08903
Availability details for
Quantity
  • Availability
  • Add to Cart