ENA024294950 Enamine

2-isopropoxy-3-methoxybenzaldehyde

  • MDL number MFCD02629693
  • Cas Number : 75792-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08911

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08911
Availability details for
Quantity
  • Availability
  • Add to Cart