ENA024294957 Enamine

3-chloro-4-ethoxy-5-methoxybenzoic acid

  • MDL number MFCD05739841
  • Cas Number : 712294-48-9
  • Molecular Weight : 230.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-08919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08919
Availability details for
Quantity
  • Availability
  • Add to Cart