ENA024294958 Enamine

6-chloro-3H-imidazo[4,5-b]pyridine-2-thiol

  • MDL number MFCD18323873
  • Cas Number : 19918-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-08920

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08920
Availability details for
Quantity
  • Availability
  • Add to Cart