ENA024295005 Enamine

3-(4-bromophenyl)-2-methyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD00437360
  • Cas Number : 1788-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-08967

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08967
Availability details for
Quantity
  • Availability
  • Add to Cart