ENA024295014 Enamine

1-(3-chlorobenzenesulfonyl)piperazine

  • MDL number MFCD05262731
  • Cas Number : 233261-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82-83
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-08976

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08976
Availability details for
Quantity
  • Availability
  • Add to Cart