ENA024295020 Enamine

5,6-dimethyl-3-(propan-2-yl)-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD05263760
  • Cas Number : 743452-35-9
  • Molecular Weight : 254.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08982

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08982
Availability details for
Quantity
  • Availability
  • Add to Cart