ENA024295030 Enamine

2-(2-fluorobenzenesulfonamido)-4-(methylsulfanyl)butanoic acid

  • MDL number MFCD03618989
  • Cas Number : 1008243-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08992

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08992
Availability details for
Quantity
  • Availability
  • Add to Cart