ENA024295041 Enamine

6-bromo-3-phenyl-2-sulfanyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD00433659
  • Cas Number : 18009-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-09003

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09003
Availability details for
Quantity
  • Availability
  • Add to Cart