ENA024295044 Enamine

3-(2,3-dimethylphenyl)-5-ethyl-6-methyl-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD05263776
  • Cas Number : 743452-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09006
Availability details for
Quantity
  • Availability
  • Add to Cart