ENA024295052 Enamine

3-(3-ethoxypropyl)-5,6-dimethyl-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD05266685
  • Cas Number : 744227-02-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-09014

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09014
Availability details for
Quantity
  • Availability
  • Add to Cart