ENA024295067 Enamine

6-ethyl-2-mercapto-3-(3-methylbutyl)thieno[2,3-d]pyrimidin-4(3H)-one

  • MDL number MFCD05267649
  • Cas Number : 744242-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09029

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09029
Availability details for
Quantity
  • Availability
  • Add to Cart