ENA024295072 Enamine

2-{[(4-methyl-1,3-thiazol-2-yl)thio]methyl}benzoic acid

  • MDL number MFCD05263788
  • Cas Number : 743453-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...162
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09034

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09034
Availability details for
Quantity
  • Availability
  • Add to Cart